accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 36094-04-9

IUPAC Name: 2-phenyl-1,3-thiazole-4-carbonyl chloride
Synonyms: 2-phenyl-1,3-thiazole-4-carbonyl chloride
Molecular Weight (g/mol): 223.67
Molecular Formula: C10H6ClNOS
InChi Key: ZZFOIDMEHXGBKS-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CSC(=N1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results