CAS RN 36099-95-3
CAS RN 36099-95-3
IUPAC Name:
2-[2-amino-3-(1H-indol-3-yl)propanamido]pentanedioic acid
Synonyms:
2-[2-amino-3-(1H-indol-3-yl)propanamido]pentanedioic acid
Molecular Weight (g/mol):
333.34
Molecular Formula:
C16H19N3O5
InChi Key:
PWIQCLSQVQBOQV-UHFFFAOYNA-N
SMILES:
NC(CC1=CNC2=CC=CC=C12)C(=O)NC(CCC(O)=O)C(O)=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals G3335
CAS: 36099-95-3 Molecular Formula: C16H19N3O5,C16H19N3O5 Synonym: H-Trp-Glu-OH