accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3610-36-4

IUPAC Name: 2-(6-methoxy-1H-indol-3-yl)ethan-1-aminium
Synonyms: 2-(6-methoxy-1H-indol-3-yl)ethanaminium
Molecular Weight (g/mol): 191.25
Molecular Formula: C11H15N2O
InChi Key: VOCGEKMEZOPDFP-UHFFFAOYSA-O
SMILES: COC1=CC=C2C(CC[NH3+])=CNC2=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results