accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CNCS

CAS RN 36176-31-5

IUPAC Name: 1-isothiocyanato-2-(propan-2-yl)benzene
Synonyms: 1-isopropyl-2-isothiocyanatobenzene
Molecular Weight (g/mol): 177.27
Molecular Formula: C10H11NS
InChi Key: QQOOMDSEMQHMIR-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC=C1N=C=S
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)