CAS RN 3619-02-1
CAS RN 3619-02-1
IUPAC Name:
methyl 2-acetamidopropanoate
Synonyms:
alanine-N-acetyl,methyl ester
Molecular Weight (g/mol):
145.16
Molecular Formula:
C6H11NO3
InChi Key:
FQGVVDYNRHNTCK-UHFFFAOYNA-N
SMILES:
COC(=O)C(C)NC(C)=O
1
–
1
of
1
results
1
N-Acetyl-L-alanine methyl ester, 95%, Thermo Scientific™
CAS: 3619-02-1 MDL Number: MFCD03940666