accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3619-22-5

IUPAC Name: 4-methylbenzohydrazide
Synonyms: benzoylhydrazine, P-methyl
Molecular Weight (g/mol): 150.18
Molecular Formula: C8H10N2O
InChi Key: MFFVZXOPRXMVET-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(=O)NN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results