accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3622-35-3

OOHSN

CAS RN 3622-35-3

IUPAC Name: 1,3-benzothiazole-6-carboxylic acid
Synonyms: 1,3-benzothiazole-6-carboxylic acid
Molecular Weight (g/mol): 179.19
Molecular Formula: C8H5NO2S
InChi Key: DMPZJACLHDWUFS-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C2N=CSC2=C1
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results