accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 36229-42-2

IUPAC Name: magnesium(2+) 3-chlorobenzen-1-ide bromide
Synonyms: magnesium(2+) 3-chlorobenzen-1-ide bromide
Molecular Weight (g/mol): 215.76
Molecular Formula: C6H4BrClMg
InChi Key: ZMPYQGQHGLLBQI-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].ClC1=C[C-]=CC=C1
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results