accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3625-57-8

IUPAC Name: 12-amino-8-(diethylamino)-10λ⁴-oxa-5-azatetraphen-10-ylium chloride
Synonyms: nile blue cresyl fast violet cresol fast violet nile blue dye nile blue chloride
Molecular Weight (g/mol): 353.85
Molecular Formula: C20H20ClN3O
InChi Key: XJCPMUIIBDVFDM-UHFFFAOYSA-M
SMILES: [Cl-].CCN(CC)C1=CC2=[O+]C3=C(N=C2C=C1)C1=CC=CC=C1C(N)=C3
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (1)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)