accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHONNHOHSOHOOSHOOO(E)

CAS RN 3627-01-8

IUPAC Name: 4-hydroxy-5-{2-[(1E)-2-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene]hydrazin-1-yl}naphthalene-2,7-disulfonic acid
Synonyms: 4-hydroxy-5-{2-[(1E)-2-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene]hydrazin-1-yl}naphthalene-2,7-disulfonic acid
Molecular Weight (g/mol): 440.4
Molecular Formula: C16H12N2O9S2
InChi Key: CYJIGBWVVZHXRF-SFQUDFHCSA-N
SMILES: OC1=CC(=O)C=C\C1=N/NC1=C2C(O)=CC(=CC2=CC(=C1)S(O)(=O)=O)S(O)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)