accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2FCl

CAS RN 363-51-9

IUPAC Name: 2-chloro-6-fluoroaniline
Synonyms: 2-chloro-6-fluoroaniline
Molecular Weight (g/mol): 145.56
Molecular Formula: C6H5ClFN
InChi Key: ZJLAWMDJTMMTQB-UHFFFAOYSA-N
SMILES: NC1=C(F)C=CC=C1Cl
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results