accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NH2FFF

CAS RN 364-13-6

IUPAC Name: 2-(trifluoromethyl)benzene-1,4-diamine
Synonyms: 2-(trifluoromethyl)benzene-1,4-diamine
Molecular Weight (g/mol): 176.14
Molecular Formula: C7H7F3N2
InChi Key: ZQQOGBKIFPCFMJ-UHFFFAOYSA-N
SMILES: NC1=CC=C(N)C(=C1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results