accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OHNOHOHHOOOOOCH3

CAS RN 36476-29-6

IUPAC Name: N-[4,5-dihydroxy-6-(hydroxymethyl)-2-[(4-methyl-2-oxo-2H-chromen-7-yl)oxy]oxan-3-yl]acetamide
Synonyms: N-[4,5-dihydroxy-6-(hydroxymethyl)-2-[(4-methyl-2-oxochromen-7-yl)oxy]oxan-3-yl]acetamide
Molecular Weight (g/mol): 379.37
Molecular Formula: C18H21NO8
InChi Key: QCTHLCFVVACBSA-UHFFFAOYNA-N
SMILES: CC(=O)NC1C(O)C(O)C(CO)OC1OC1=CC=C2C(C)=CC(=O)OC2=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)