accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 36520-39-5

IUPAC Name: azetidine hydrochloride
Synonyms: azetidine hydrochloride
Molecular Weight (g/mol): 93.55
Molecular Formula: C3H8ClN
InChi Key: HGQULGDOROIPJN-UHFFFAOYSA-N
SMILES: Cl.C1CNC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)