accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2FFCl

CAS RN 36556-56-6

IUPAC Name: 2-chloro-4,6-difluoroaniline
Synonyms: 2-chloro-4,6-difluoroaniline
Molecular Weight (g/mol): 163.55
Molecular Formula: C6H4ClF2N
InChi Key: DUPRZIYJXCCXQZ-UHFFFAOYSA-N
SMILES: NC1=C(F)C=C(F)C=C1Cl
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)