accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BOOH3CCH3H3CH3CNSiCH3CH3CH3H3CCH3H3C

CAS RN 365564-11-0

IUPAC Name: 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-[tris(propan-2-yl)silyl]-1H-pyrrole
Synonyms: 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-(triisopropylsilyl)pyrrole
Molecular Weight (g/mol): 349.4
Molecular Formula: C19H36BNO2Si
InChi Key: GWFIZBYDIHGZRJ-UHFFFAOYSA-N
SMILES: CC(C)[Si](C(C)C)(C(C)C)N1C=CC(=C1)B1OC(C)(C)C(C)(C)O1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results