accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OH(S)OO

CAS RN 36567-72-3

IUPAC Name: (3S)-3-hydroxy-3-phenylpropanoate
Synonyms: (3S)-3-hydroxy-3-phenylpropanoate
Molecular Weight (g/mol): 165.17
Molecular Formula: C9H9O3
InChi Key: AYOLELPCNDVZKZ-QMMMGPOBSA-M
SMILES: O[C@@H](CC([O-])=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results