accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaNaOSOOSOOO

CAS RN 36589-61-4

IUPAC Name: disodium butane-1,4-disulfonate
Synonyms: disodium butane-1,4-disulfonate
Molecular Weight (g/mol): 262.2
Molecular Formula: C4H8Na2O6S2
InChi Key: LEUIUWYZAHKPSE-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)CCCCS([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results