CAS RN 36592-77-5
CAS RN 36592-77-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Metipranolol Hydrochloride, TRC
CAS: 36592-77-5 Molecular Formula: C17H28ClNO4 Molecular Weight (g/mol): 345.86 Synonym: Betamann,Optipranolol,1-(2,3,5-Trimethyl-4-acetoxyphenoxy)-3(isopropylamino)-2-propanol monohydro chloride,4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]-2,3,6-trimethylphenol 1-Acetate Hydrochloride IUPAC Name: 4-(2-hydroxy-3-(isopropylamino)propoxy)-2,3,6-trimethylphenyl acetate hydrochloride SMILES: O=C(C)OC1=C(C)C(C)=C(OCC(O)CNC(C)C)C=C1C.Cl