accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 365996-06-1

IUPAC Name: tert-butyl 2-bromo-4H,5H,6H,7H-[1,3]thiazolo[5,4-c]pyridine-5-carboxylate
Synonyms: tert-butyl 2-bromo-4H,6H,7H-[1,3]thiazolo[5,4-c]pyridine-5-carboxylate
Molecular Weight (g/mol): 319.22
Molecular Formula: C11H15BrN2O2S
InChi Key: RXXKNZKFRAFFOH-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC2=C(C1)SC(Br)=N2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results