accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 366-70-1

IUPAC Name: hydrogen 4-[(2-methylhydrazin-1-yl)methyl]-N-(propan-2-yl)benzamide chloride
Synonyms: hydrogen procarbazine chloride
Molecular Weight (g/mol): 257.76
Molecular Formula: C12H20ClN3O
InChi Key: DERJYEZSLHIUKF-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CNNCC1=CC=C(C=C1)C(=O)NC(C)C
Quantity 
  • (2)
  • (1)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)
Percent Purity 
  • (2)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results