CAS RN 366017-09-6
IUPAC Name:
1-{4-[4-({2-[(1E)-2-[4-(trifluoromethyl)phenyl]ethenyl]-1,3-oxazol-4-yl}methoxy)phenyl]butyl}-1H-1,2,3-triazole
Synonyms:
mubritinib
Molecular Weight (g/mol):
468.48
Molecular Formula:
C25H23F3N4O2
InChi Key:
ZTFBIUXIQYRUNT-MDWZMJQESA-N
SMILES:
FC(F)(F)C1=CC=C(\C=C\C2=NC(COC3=CC=C(CCCCN4C=CN=N4)C=C3)=CO2)C=C1