CAS RN 36635-61-7
IUPAC Name:
1-isocyanomethanesulfonyl-4-methylbenzene
Synonyms:
TosMIC
Molecular Weight (g/mol):
195.24
Molecular Formula:
C9H9NO2S
InChi Key:
CFOAUYCPAUGDFF-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)S(=O)(=O)C[N+]#[C-]
1
–
4
of
4
results