accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOMgBr

CAS RN 36637-44-2

IUPAC Name: magnesium(2+) 4-(oxan-2-yloxy)benzen-1-ide bromide
Synonyms: magnesium(2+) 4-(oxan-2-yloxy)benzen-1-ide bromide
Molecular Weight (g/mol): 281.43
Molecular Formula: C11H13BrMgO2
InChi Key: JNWNAQZSBRFGPC-UHFFFAOYNA-M
SMILES: [Mg++].[Br-].C1CCC(OC2=CC=[C-]C=C2)OC1
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)