accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FF

CAS RN 367-11-3

IUPAC Name: 1,2-difluorobenzene
Synonyms: 1,2-difluorobenzene
Molecular Weight (g/mol): 114.1
Molecular Formula: C6H4F2
InChi Key: GOYDNIKZWGIXJT-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1F
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)