accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2FCl

CAS RN 367-21-5

IUPAC Name: 3-chloro-4-fluoroaniline
Synonyms: 3-chloro-4-fluoroaniline
Molecular Weight (g/mol): 145.56
Molecular Formula: C6H5ClFN
InChi Key: YSEMCVGMNUUNRK-UHFFFAOYSA-N
SMILES: NC1=CC=C(F)C(Cl)=C1
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results