accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3FH2N

CAS RN 367-29-3

IUPAC Name: 5-fluoro-2-methylaniline
Synonyms: 5-fluoro-2-methylaniline
Molecular Weight (g/mol): 125.15
Molecular Formula: C7H8FN
InChi Key: JLCDTNNLXUMYFQ-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C=C1N
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)