CAS RN 367-93-1
Isopropyl-beta-D-thiogalactoside, dioxane-free, 99%, Thermo Scientific Chemicals
CAS: 367-93-1 Molecular Formula: C9H18O5S Molecular Weight (g/mol): 238.30 MDL Number: MFCD00063273 InChI Key: BPHPUYQFMNQIOC-NXRLNHOXSA-N Synonym: iptg,isopropyl-beta-d-thiogalactopyranoside,isopropyl-beta-d-thiogalactoside,isopropyl thiogalactoside,isopropyl beta-d-thiogalactopyranoside,isopropyl beta-d-thiogalactoside,isopropyl beta-d-1-thiogalactopyranoside,unii-x73vv2246b,2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol,isopropyl-1-beta-d-thiogalactoside PubChem CID: 656894 ChEBI: CHEBI:61448 IUPAC Name: (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-propan-2-ylsulfanyloxane-3,4,5-triol SMILES: CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
Thermo Scientific Chemicals Isopropyl-beta-D-thiogalactopyranoside, 99%, for biochemistry, dioxane-free
CAS: 367-93-1 Molecular Formula: C9H18O5S Molecular Weight (g/mol): 238.30 MDL Number: MFCD00063273 InChI Key: BPHPUYQFMNQIOC-NXRLNHOXSA-N Synonym: iptg,isopropyl-beta-d-thiogalactopyranoside,isopropyl-beta-d-thiogalactoside,isopropyl thiogalactoside,isopropyl beta-d-thiogalactopyranoside,isopropyl beta-d-thiogalactoside,isopropyl beta-d-1-thiogalactopyranoside,unii-x73vv2246b,2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol,isopropyl-1-beta-d-thiogalactoside PubChem CID: 656894 ChEBI: CHEBI:61448 IUPAC Name: (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-propan-2-ylsulfanyloxane-3,4,5-triol SMILES: CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
Isopropyl-beta-D-thiogalactopyranoside (IPTG), Dioxane free, >99%, Ultrapure, Thermo Scientific Chemicals
CAS: 367-93-1 Molecular Formula: C9H18O5S Molecular Weight (g/mol): 238.298 MDL Number: MFCD00063273 InChI Key: BPHPUYQFMNQIOC-NXRLNHOXSA-N Synonym: iptg,isopropyl-beta-d-thiogalactopyranoside,isopropyl-beta-d-thiogalactoside,isopropyl thiogalactoside,isopropyl beta-d-thiogalactopyranoside,isopropyl beta-d-thiogalactoside,isopropyl beta-d-1-thiogalactopyranoside,unii-x73vv2246b,2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol,isopropyl-1-beta-d-thiogalactoside PubChem CID: 656894 ChEBI: CHEBI:61448 IUPAC Name: (2 R,3 R,4 S,5 R,6 S)-2-(hydroxymethyl)-6-propan-2-ylsulfanyloxane-3,4,5-triol SMILES: CC(C)SC1C(C(C(C(O1)CO)O)O)O
Isopropyl-β-D-thiogalactopyranoside, MP Biomedicals™
CAS: 367-93-1 Molecular Formula: C9H18O5S Molecular Weight (g/mol): 238.30 MDL Number: MFCD00063273 InChI Key: BPHPUYQFMNQIOC-NXRLNHOXSA-N Synonym: IPTG,Isopropyl β-D-1-thiogalactopyranoside IUPAC Name: (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol SMILES: CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
Isopropyl-β-D-Thiogalactopyranoside, (Crystalline),MP Biomedicals
CAS: 367-93-1 Molecular Formula: C9H18O5S Molecular Weight (g/mol): 238.30 MDL Number: MFCD00063273 InChI Key: BPHPUYQFMNQIOC-NXRLNHOXSA-N Synonym: iptg,isopropyl-beta-d-thiogalactopyranoside,isopropyl-beta-d-thiogalactoside,isopropyl thiogalactoside,isopropyl beta-d-thiogalactopyranoside,isopropyl beta-d-thiogalactoside,isopropyl beta-d-1-thiogalactopyranoside,unii-x73vv2246b,2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol,isopropyl-1-beta-d-thiogalactoside PubChem CID: 656894 ChEBI: CHEBI:61448 SMILES: CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
Isopropyl-β-D-Thiogalactopyranoside, >99%, MP Biomedicals
CAS: 367-93-1 Molecular Formula: C9H18O5S Molecular Weight (g/mol): 238.30 MDL Number: MFCD00063273 InChI Key: BPHPUYQFMNQIOC-NXRLNHOXSA-N Synonym: iptg,isopropyl-beta-d-thiogalactopyranoside,isopropyl-beta-d-thiogalactoside,isopropyl thiogalactoside,isopropyl beta-d-thiogalactopyranoside,isopropyl beta-d-thiogalactoside,isopropyl beta-d-1-thiogalactopyranoside,unii-x73vv2246b,2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol,isopropyl-1-beta-d-thiogalactoside PubChem CID: 656894 ChEBI: CHEBI:61448 SMILES: CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
Isopropyl ß-D-Thiogalactopyranoside, Dioxane Free, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
R&D Systems™ IPTG
A Tocris product, part of R&D Systems. Used in cloning procedures with X-GAL
Melford IPTG, Dioxane Free, Non-Mammalian
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
Melford IPTG
IPTG is combined with X-GAL to detect lac+ colonies or cells in a colorimetric assay.
Isopropyl ß-D-Thiogalactopyranoside, Dioxane Free, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Isopropyl-beta-D-thiogalactopyranoside, 99+%, for molecular biology, DNAse, RNAse an, Thermo Scientific™
CAS: 367-93-1 Molecular Formula: C9H18O5S Molecular Weight (g/mol): 238.30 MDL Number: MFCD00063273 InChI Key: BPHPUYQFMNQIOC-NXRLNHOXSA-N Synonym: iptg,isopropyl-beta-d-thiogalactopyranoside,isopropyl-beta-d-thiogalactoside,isopropyl thiogalactoside,isopropyl beta-d-thiogalactopyranoside,isopropyl beta-d-thiogalactoside,isopropyl beta-d-1-thiogalactopyranoside,unii-x73vv2246b,2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol,isopropyl-1-beta-d-thiogalactoside PubChem CID: 656894 ChEBI: CHEBI:61448 IUPAC Name: (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-propan-2-ylsulfanyloxane-3,4,5-triol SMILES: CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
Isopropyl-β-D-thiogalactopyranoside (IPTG) (Dioxane-free), Fisher BioReagents™
CAS: 367-93-1 Molecular Formula: C9H18O5S Molecular Weight (g/mol): 238.30 MDL Number: MFCD00063273 InChI Key: BPHPUYQFMNQIOC-NXRLNHOXSA-N Synonym: iptg,isopropyl-beta-d-thiogalactopyranoside,isopropyl-beta-d-thiogalactoside,isopropyl thiogalactoside,isopropyl beta-d-thiogalactopyranoside,isopropyl beta-d-thiogalactoside,isopropyl beta-d-1-thiogalactopyranoside,unii-x73vv2246b,2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol,isopropyl-1-beta-d-thiogalactoside PubChem CID: 656894 ChEBI: CHEBI:61448 SMILES: CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
Isopropyl-β-D-thiogalactopyranoside (IPTG) (Wht. Powd., Dioxane Crystalline), Fisher BioReagents™
CAS: 367-93-1 Molecular Formula: C9H18O5S Molecular Weight (g/mol): 238.30 MDL Number: MFCD00063273 InChI Key: BPHPUYQFMNQIOC-NXRLNHOXSA-N Synonym: dioxane,p-dioxane,1,4-diethylene dioxide,diethylene ether,dioxan,1,4-dioxacyclohexane,diethylene dioxide,dioxanne,di ethylene oxide,tetrahydro-p-dioxin PubChem CID: 31275 ChEBI: CHEBI:47032 IUPAC Name: 1,4-dioxane SMILES: CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
- 1
- 2