CAS RN 36734-19-7
CAS RN 36734-19-7
IUPAC Name:
3-(3,5-dichlorophenyl)-2,4-dioxo-N-(propan-2-yl)imidazolidine-1-carboxamide
Synonyms:
iprodione rovral rovral (iprodione)
Molecular Weight (g/mol):
330.17
Molecular Formula:
C13H13Cl2N3O3
InChi Key:
ONUFESLQCSAYKA-UHFFFAOYSA-N
SMILES:
CC(C)NC(=O)N1CC(=O)N(C1=O)C1=CC(Cl)=CC(Cl)=C1
1
–
9
of
9
results
2
Iprodione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Iprodione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Iprodione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
5
Iprodione 1000 μg/mL in Acetone, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality