accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2OHNO

CAS RN 3676-85-5

IUPAC Name: 5-amino-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 5-amino-2H-isoindole-1,3-dione
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: PXRKCOCTEMYUEG-UHFFFAOYSA-N
SMILES: NC1=CC=C2C(=O)NC(=O)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results