accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 36768-62-4

CH3H3CNH3H3CH3CH2N

CAS RN 36768-62-4

IUPAC Name: 4-azaniumyl-2,2,6,6-tetramethylpiperidin-1-ium
Synonyms: 4-ammonio-2,2,6,6-tetramethylpiperidin-1-ium
Molecular Weight (g/mol): 158.29
Molecular Formula: C9H22N2
InChi Key: FTVFPPFZRRKJIH-UHFFFAOYSA-P
SMILES: CC1(C)CC([NH3+])CC(C)(C)[NH2+]1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results