accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 36770-50-0

IUPAC Name: 2-[3-(pyridin-4-yl)-1H-1,2,4-triazol-5-yl]pyridine
Synonyms: 2-[5-(pyridin-4-yl)-2H-1,2,4-triazol-3-yl]pyridine
Molecular Weight (g/mol): 223.24
Molecular Formula: C12H9N5
InChi Key: WOFXUCJTFGKKIF-UHFFFAOYSA-N
SMILES: N1N=C(N=C1C1=CC=CC=N1)C1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results