CAS RN 3678-63-5
IUPAC Name:
4-chloro-2-methylpyridine
Synonyms:
4-chloro-2-methylpyridine
Molecular Weight (g/mol):
127.57
Molecular Formula:
C6H6ClN
InChi Key:
DAOZBJCTEPJGES-UHFFFAOYSA-N
SMILES:
CC1=NC=CC(Cl)=C1
1
–
1
of
1
results
1
4-Chloro-2-methylpyridine, 95%
CAS: 3678-63-5 Molecular Formula: C6H6ClN Molecular Weight (g/mol): 127.571 MDL Number: MFCD00038833 InChI Key: DAOZBJCTEPJGES-UHFFFAOYSA-N Synonym: 4-chloro-2-picoline,2-methyl-4-chloropyridine,4-chloro-2-methyl-pyridine,pyridine, 4-chloro-2-methyl,p-chloropicoline,4-chloropicoline,4-chloro-2-picolin,acmc-20amln,pubchem2071 PubChem CID: 581392 IUPAC Name: 4-chloro-2-methylpyridine SMILES: CC1=NC=CC(=C1)Cl