accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2ONNN(R)O(R)OH(S)HO(R)HO

CAS RN 36791-04-5

IUPAC Name: 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1H-1,2,4-triazole-3-carboxamide
Synonyms: ribavirin viramid ribavirin-TP virazid ribavirin triphosphate copegus 利巴韦林 grossman brand of ribavirin rebetol rebetron
Molecular Weight (g/mol): 244.21
Molecular Formula: C8H12N4O5
InChi Key: IWUCXVSUMQZMFG-AFCXAGJDSA-N
SMILES: NC(=O)C1=NN(C=N1)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O
Molecular Weight (g/mol) 
  • (9)
  • (1)
Percent Purity 
  • (3)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (9)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)