accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 368-71-8

NH2H2NFFF

CAS RN 368-71-8

IUPAC Name: 4-(trifluoromethyl)benzene-1,2-diamine
Synonyms: 4-(trifluoromethyl)benzene-1,2-diamine
Molecular Weight (g/mol): 176.14
Molecular Formula: C7H7F3N2
InChi Key: RQWJHUJJBYMJMN-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1N)C(F)(F)F
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (3)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)