accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NHH3COH3CCH3H3COCH3CH3H3C

CAS RN 36805-97-7

IUPAC Name: [bis(tert-butoxy)methyl]dimethylazanium
Synonyms: [bis(tert-butoxy)methyl]dimethylazanium
Molecular Weight (g/mol): 204.33
Molecular Formula: C11H26NO2
InChi Key: DBNQIOANXZVWIP-UHFFFAOYSA-O
SMILES: C[NH+](C)C(OC(C)(C)C)OC(C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)