accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OH(R)O(S)(R)OH(R)OH(S)OHHOOOOH

CAS RN 3681-99-0

IUPAC Name: 7-hydroxy-3-(4-hydroxyphenyl)-8-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-4H-chromen-4-one
Synonyms: puerarin
Molecular Weight (g/mol): 416.38
Molecular Formula: C21H20O9
InChi Key: HKEAFJYKMMKDOR-VPRICQMDSA-N
SMILES: OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)C1=C2OC=C(C(=O)C2=CC=C1O)C1=CC=C(O)C=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)