accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NONHNHO

CAS RN 3682-14-2

IUPAC Name: 6-amino-1,2,3,4-tetrahydrophthalazine-1,4-dione
Synonyms: 4-aminophthalhydrazide
Molecular Weight (g/mol): 177.16
Molecular Formula: C8H7N3O2
InChi Key: HUDPLKWXRLNSPC-UHFFFAOYSA-N
SMILES: NC1=CC=C2C(=O)NNC(=O)C2=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results