accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNBrBr

CAS RN 36847-10-6

IUPAC Name: 4,6-dibromopyrimidine
Synonyms: 4,6-dibromopyrimidine
Molecular Weight (g/mol): 237.88
Molecular Formula: C4H2Br2N2
InChi Key: CEPVZSXIONCAJV-UHFFFAOYSA-N
SMILES: BrC1=CC(Br)=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results