accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 36847-86-6

OOONHBr(Z)

CAS RN 36847-86-6

IUPAC Name: (2Z)-3-[(4-bromophenyl)carbamoyl]prop-2-enoate
Synonyms: (2Z)-3-[(4-bromophenyl)carbamoyl]prop-2-enoate
Molecular Weight (g/mol): 269.08
Molecular Formula: C10H7BrNO3
InChi Key: DSNFUZCLEZJCRN-WAYWQWQTSA-M
SMILES: [O-]C(=O)\C=C/C(=O)NC1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results