accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2OHO

CAS RN 3685-23-2

IUPAC Name: 4-aminocyclohexane-1-carboxylic acid
Synonyms: 4-aminocyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 143.19
Molecular Formula: C7H13NO2
InChi Key: DRNGLYHKYPNTEA-UHFFFAOYSA-N
SMILES: NC1CCC(CC1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)