accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 36881-42-2

IUPAC Name: 1-(bromomethyl)-4-phenoxybenzene
Synonyms: 1-(bromomethyl)-4-phenoxybenzene
Molecular Weight (g/mol): 263.13
Molecular Formula: C13H11BrO
InChi Key: CPIGBCFBFZSCQI-UHFFFAOYSA-N
SMILES: BrCC1=CC=C(OC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results