CAS RN 368869-96-9
IUPAC Name:
3-(4-bromophenoxy)-6-methylpyridazine
Synonyms:
3-(4-bromophenoxy)-6-methylpyridazine
Molecular Weight (g/mol):
265.11
Molecular Formula:
C11H9BrN2O
InChi Key:
LZEJOINDIKGWQJ-UHFFFAOYSA-N
SMILES:
CC1=NN=C(OC2=CC=C(Br)C=C2)C=C1
1
–
1
of
1
results
1
3-(4-Bromophenoxy)-6-methylpyridazine, 97%, Thermo Scientific™
CAS: 368869-96-9 Molecular Formula: C11H9BrN2O Molecular Weight (g/mol): 265.11 MDL Number: MFCD03086135 InChI Key: LZEJOINDIKGWQJ-UHFFFAOYSA-N Synonym: 3-4-bromophenoxy-6-methylpyridazine,pyridazine,3-4-bromophenoxy-6-methyl,3-4-bromanylphenoxy-6-methyl-pyridazine,4-bromo-1-6-methylpyridazin-3-yloxy benzene PubChem CID: 2776522 IUPAC Name: 3-(4-bromophenoxy)-6-methylpyridazine SMILES: CC1=NN=C(OC2=CC=C(Br)C=C2)C=C1