accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 369-30-2

IUPAC Name: methyl 3-fluoro-4-methoxybenzoate
Synonyms: methyl 3-fluoro-4-methoxybenzoate
Molecular Weight (g/mol): 184.17
Molecular Formula: C9H9FO3
InChi Key: MIKOKYAREMINFF-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(F)=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results