accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 369-36-8

IUPAC Name: 2-fluoro-5-nitroaniline
Synonyms: 2-fluoro-5-nitroaniline
Molecular Weight (g/mol): 156.12
Molecular Formula: C6H5FN2O2
InChi Key: KJVBJICWGQIMOZ-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=C1F)[N+]([O-])=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results