accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 369-77-7

IUPAC Name: 3-[4-chloro-3-(trifluoromethyl)phenyl]-1-(4-chlorophenyl)urea
Synonyms: cloflucarban 卤卡班
Molecular Weight (g/mol): 349.13
Molecular Formula: C14H9Cl2F3N2O
InChi Key: ZFSXZJXLKAJIGS-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(NC(=O)NC2=CC=C(Cl)C=C2)=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results