accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NCS

CAS RN 3694-46-0

IUPAC Name: 1-(isothiocyanatomethyl)-4-methylbenzene
Synonyms: 1-(isothiocyanatomethyl)-4-methylbenzene
Molecular Weight (g/mol): 163.24
Molecular Formula: C9H9NS
InChi Key: OAXIUBJXQISJEV-UHFFFAOYSA-N
SMILES: CC1=CC=C(CN=C=S)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results