accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3696-22-8

IUPAC Name: (4-nitrophenyl)thiourea
Synonyms: thiourea, 3-(nitrophenyl)-
Molecular Weight (g/mol): 197.21
Molecular Formula: C7H7N3O2S
InChi Key: BLYAANPIHFKKMQ-UHFFFAOYSA-N
SMILES: NC(=S)NC1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results