accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3699-66-9

CH3OOCH3POOH3COH3C

CAS RN 3699-66-9

IUPAC Name: ethyl 2-(diethoxyphosphoryl)propanoate
Synonyms: ethyl 2-(diethoxyphosphoryl)propanoate
Molecular Weight (g/mol): 238.22
Molecular Formula: C9H19O5P
InChi Key: BVSRWCMAJISCTD-UHFFFAOYNA-N
SMILES: CCOC(=O)C(C)P(=O)(OCC)OCC
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (4)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (4)
  • (2)
  • (2)
  • (2)