accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 37025-55-1

H3C(S)CH3(S)HNO(S)HONCH3OS(R)HNO(S)NH2OHNO(S)NH2ONHOON(S)ONH(S)CH3H3COHNNH2O

CAS RN 37025-55-1

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)